Assay Details
| Gene Symbol: LOC4344057 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791848.2 | XP_015647334.1 | 2 - 3 | 833 | 104 | |
| GenBank mRNA | AK105489.1 | - | 1 - 2 | 171 | 104 | |
More Information
Set Membership: |
|

LOC4344057
Genomic Map