Assay Details
| Gene Symbol: LOC4344057 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791848.2 | XP_015647334.1 | 6 - 7 | 1438 | 144 | |
| GenBank mRNA | AK105489.1 | - | 5 - 6 | 776 | 144 | |
More Information
Set Membership: |
|

LOC4344057
Genomic Map