Assay Details
| Gene Symbol: LOC4344112 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015789119.2 | XP_015644605.1 | 3 - 4 | 442 | 70 | |
| XM_015789120.2 | XP_015644606.1 | 3 - 4 | 442 | 70 | ||
| GenBank mRNA | AK102332.1 | - | 3 - 4 | 443 | 70 | |
More Information
Set Membership: |
|

LOC4344112
Genomic Map