Assay Details
| Gene Symbol: LOC4344112 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015789119.2 | XP_015644605.1 | 6 - 7 | 739 | 113 | |
| XM_015789120.2 | XP_015644606.1 | 6 - 7 | 739 | 113 | ||
| GenBank mRNA | AK102332.1 | - | 6 - 7 | 740 | 113 | |
| AK111992.1 | - | 2 - 3 | 83 | 113 | ||
More Information
Set Membership: |
|

LOC4344112
Genomic Map