Assay Details
| Gene Symbol: LOC4344112 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015789119.2 | XP_015644605.1 | 10 - 11 | 1116 | 76 | |
| XM_015789120.2 | XP_015644606.1 | 10 - 11 | 1116 | 76 | ||
| GenBank mRNA | AK102332.1 | - | 9 - 10 | 1520 | 76 | |
| AK111992.1 | - | 6 - 7 | 460 | 76 | ||
More Information
Set Membership: |
|

LOC4344112
Genomic Map