Assay Details
Gene Symbol: LOC4344217 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015789089.2 | XP_015644575.1 | 8 - 9 | 935 | 117 | |
GenBank mRNA | AB021878.1 | - | 7 - 8 | 1035 | 117 | |
AK064004.1 | - | 8 - 9 | 925 | 117 | ||
AK099332.1 | - | 8 - 9 | 917 | 117 | ||
AY324877.1 | - | 7 - 8 | 801 | 117 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
