Assay Details
| Gene Symbol: LOC4344290 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791288.2 | XP_015646774.1 | 14 - 15 | 1261 | 96 | |
| GenBank mRNA | AK099229.1 | - | 14 - 15 | 1256 | 96 | |
| Gene Symbol: LOC4338886 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK287702.1 | - | 2025 | 96 | ||
More Information
Set Membership: |
|

LOC4344290
Genomic Map