Assay Details
| Gene Symbol: LOC4344290 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791288.2 | XP_015646774.1 | 3 - 4 | 405 | 120 | |
| GenBank mRNA | AK065079.1 | - | 3 - 4 | 400 | 120 | |
| AK099229.1 | - | 3 - 4 | 400 | 120 | ||
More Information
Set Membership: |
|

LOC4344290
Genomic Map