Assay Details
| Gene Symbol: LOC4344396 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791069.2 | XP_015646555.1 | 3 - 4 | 660 | 94 | |
| GenBank mRNA | AB300404.1 | - | 3 - 4 | 339 | 94 | |
| AK066428.1 | - | 3 - 4 | 449 | 94 | ||
More Information
Set Membership: |
|

LOC4344396
Genomic Map