Assay Details
| Gene Symbol: LOC4344663 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015792955.2 | XP_015648441.1 | 4 - 5 | 828 | 96 | |
| GenBank mRNA | AK101530.1 | - | 4 - 5 | 781 | 96 | |
More Information
Set Membership: |
|

LOC4344663
Genomic Map