Assay Details
| Gene Symbol: LOC4344697 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015795331.2 | XP_015650817.1 | 2 - 3 | 394 | 64 | |
| GenBank mRNA | AB219538.1 | - | 2 - 3 | 309 | 64 | |
| AK073344.1 | - | 2 - 3 | 385 | 64 | ||
| AK106397.1 | - | 2 - 3 | 304 | 64 | ||
More Information
Set Membership: |
|

LOC4344697
Genomic Map