Assay Details
| Gene Symbol: LOC4344697 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015795331.2 | XP_015650817.1 | 3 - 4 | 531 | 86 | |
| GenBank mRNA | AB219538.1 | - | 3 - 4 | 446 | 86 | |
| AK106397.1 | - | 3 - 4 | 441 | 86 | ||
More Information
Set Membership: |
|

LOC4344697
Genomic Map