Assay Details
| Gene Symbol: LOC4344832 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015794536.2 | XP_015650022.1 | 1 - 2 | 224 | 71 | |
| XM_015794537.2 | XP_015650023.1 | 1 - 2 | 224 | 71 | ||
| GenBank mRNA | AK120496.1 | - | 1 - 2 | 194 | 71 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015794536.2, XM_015794537.2 transcripts.
Set Membership: |
|

LOC4344832
Genomic Map