Assay Details
| Gene Symbol: LOC4344832 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015794536.2 | XP_015650022.1 | 6 - 7 | 1679 | 118 | |
| XM_015794537.2 | XP_015650023.1 | 6 - 7 | 1679 | 118 | ||
| GenBank mRNA | AK120496.1 | - | 6 - 7 | 1635 | 118 | |
More Information
Set Membership: |
|

LOC4344832
Genomic Map