Assay Details
| Gene Symbol: LOC4344928 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015792818.1 | XP_015648304.1 | 12 - 13 | 1009 | 84 | |
| GenBank mRNA | AK103274.1 | - | 12 - 13 | 1021 | 84 | |
More Information
Set Membership: |
|

LOC4344928
Genomic Map