Assay Details
| Gene Symbol: LOC4344928 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015792818.1 | XP_015648304.1 | 1 - 2 | 138 | 73 | |
| GenBank mRNA | AK103274.1 | - | 1 - 2 | 150 | 73 | |
More Information
Set Membership: |
|

LOC4344928
Genomic Map