Assay Details
| Gene Symbol: LOC4345097 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015795409.2 | XP_015650895.1 | 4 - 5 | 1087 | 128 | |
| GenBank mRNA | AK072225.1 | - | 4 - 5 | 1083 | 128 | |
| AK121121.1 | - | 4 - 5 | 1060 | 128 | ||
More Information
Set Membership: |
|

LOC4345097
Genomic Map