Assay Details
| Gene Symbol: LOC4345097 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015795409.2 | XP_015650895.1 | 8 - 9 | 2703 | 126 | |
| GenBank mRNA | AK121121.1 | - | 8 - 9 | 2676 | 126 | |
More Information
Set Membership: |
|

LOC4345097
Genomic Map