Assay Details
| Gene Symbol: LOC4345252 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015794774.2 | XP_015650260.1 | 1 - 2 | 501 | 131 | |
| XM_015794775.2 | XP_015650261.1 | 1 - 2 | 504 | 131 | ||
| GenBank mRNA | AK108030.1 | - | 1 - 2 | 502 | 131 | |
More Information
Set Membership: |
|

LOC4345252
Genomic Map