Assay Details
| Gene Symbol: LOC4345252 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015794774.2 | XP_015650260.1 | 3 - 4 | 617 | 99 | |
| XM_015794775.2 | XP_015650261.1 | 3 - 4 | 620 | 99 | ||
| GenBank mRNA | AK108030.1 | - | 3 - 4 | 618 | 99 | |
More Information
Set Membership: |
|

LOC4345252
Genomic Map