Assay Details
| Gene Symbol: LOC4345455 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015794912.2 | XP_015650398.1 | 1 - 2 | 160 | 122 | |
| GenBank mRNA | AK070807.1 | - | 1 - 2 | 160 | 122 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015794912.2 transcripts.
Set Membership: |
|

LOC4345455
Genomic Map