Assay Details
| Gene Symbol: LOC4345455 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015794912.2 | XP_015650398.1 | 7 - 8 | 760 | 69 | |
| GenBank mRNA | AK070807.1 | - | 7 - 8 | 760 | 69 | |
More Information
Set Membership: |
|

LOC4345455
Genomic Map