Assay Details
| Gene Symbol: LOC4345700 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015794102.2 | XP_015649588.1 | 1 | 942 | 131 | |
| GenBank mRNA | AK110520.1 | - | 1 | 948 | 131 | |
More Information
Set Membership: |
|

LOC4345700
Genomic Map