Assay Details
| Gene Symbol: LOC4346248 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015795190.2 | XP_015650676.1 | 12 - 13 | 2101 | 67 | |
| GenBank mRNA | AK070916.1 | - | 8 - 9 | 1112 | 67 | |
More Information
Set Membership: |
|

LOC4346248
Genomic Map