Assay Details
| Gene Symbol: LOC4346248 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015795190.2 | XP_015650676.1 | 5 - 6 | 1127 | 59 | |
| GenBank mRNA | AK069066.1 | - | 5 - 6 | 761 | 59 | |
| AK070916.1 | - | 1 - 2 | 138 | 59 | ||
More Information
Set Membership: |
|

LOC4346248
Genomic Map