Assay Details
| Gene Symbol: LOC4346248 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015795190.2 | XP_015650676.1 | 6 - 7 | 1224 | 65 | |
| GenBank mRNA | AK069066.1 | - | 6 - 7 | 858 | 65 | |
| AK070916.1 | - | 2 - 3 | 235 | 65 | ||
More Information
Set Membership: |
|

LOC4346248
Genomic Map