Assay Details
| Gene Symbol: LOC4346287 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015792810.2 | XP_015648296.1 | 4 - 5 | 930 | 144 | |
| GenBank mRNA | AK099715.1 | - | 4 - 5 | 754 | 144 | |
More Information
Set Membership: |
|

LOC4346287
Genomic Map