Assay Details
| Gene Symbol: LOC4346342 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015793626.2 | XP_015649112.1 | 1 | 1748 | 105 | |
| GenBank mRNA | AK119787.1 | - | 1 | 1754 | 105 | |
More Information
Set Membership: |
|

LOC4346342
Genomic Map