Assay Details
| Gene Symbol: LOC4347315 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015755368.2 | XP_015610854.1 | 1 | 1399 | 89 | |
| GenBank mRNA | AK108246.1 | - | 1 | 957 | 89 | |
More Information
Set Membership: |
|

LOC4347315
Genomic Map