Assay Details
| Gene Symbol: LOC4347318 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015756224.2 | XP_015611710.1 | 1 - 2 | 415 | 81 | |
| GenBank mRNA | AK063444.1 | - | 1 - 2 | 347 | 81 | |
More Information
Set Membership: |
|

LOC4347318
Genomic Map