Assay Details
| Gene Symbol: LOC4347359 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015757038.2 | XP_015612524.1 | 11 - 12 | 1639 | 83 | |
| XM_015757039.2 | XP_015612525.1 | 10 - 11 | 1385 | 83 | ||
| GenBank mRNA | AK071315.1 | - | 11 - 12 | 1615 | 83 | |
| AK101896.1 | - | 9 - 10 | 2882 | 83 | ||
| EU837261.1 | - | 11 - 12 | 1608 | 83 | ||
More Information
Set Membership: |
|

LOC4347359
Genomic Map