Assay Details
| Gene Symbol: LOC4347359 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015757038.2 | XP_015612524.1 | 1 - 2 | 260 | 84 | |
| XM_015757039.2 | XP_015612525.1 | 1 - 2 | 258 | 84 | ||
| GenBank mRNA | AK071315.1 | - | 1 - 2 | 236 | 84 | |
| EU837261.1 | - | 1 - 2 | 229 | 84 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015757038.2, XM_015757039.2 transcripts.
Set Membership: |
|

LOC4347359
Genomic Map