Assay Details
| Gene Symbol: LOC4347359 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015757038.2 | XP_015612524.1 | 5 - 6 | 949 | 93 | |
| XM_015757039.2 | XP_015612525.1 | 4 - 5 | 695 | 93 | ||
| GenBank mRNA | AK071315.1 | - | 5 - 6 | 925 | 93 | |
| AK101896.1 | - | 3 - 4 | 2192 | 93 | ||
| EU837261.1 | - | 5 - 6 | 918 | 93 | ||
More Information
Set Membership: |
|

LOC4347359
Genomic Map