Assay Details
| Gene Symbol: LOC4347359 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015757038.2 | XP_015612524.1 | 6 - 7 | 1018 | 102 | |
| XM_015757039.2 | XP_015612525.1 | 5 - 6 | 764 | 102 | ||
| GenBank mRNA | AK071315.1 | - | 6 - 7 | 994 | 102 | |
| AK101896.1 | - | 4 - 5 | 2261 | 102 | ||
| EU837261.1 | - | 6 - 7 | 987 | 102 | ||
More Information
Set Membership: |
|

LOC4347359
Genomic Map