Assay Details
| Gene Symbol: LOC4347359 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015757038.2 | XP_015612524.1 | 8 - 9 | 1160 | 109 | |
| XM_015757039.2 | XP_015612525.1 | 7 - 8 | 906 | 109 | ||
| GenBank mRNA | AK071315.1 | - | 8 - 9 | 1136 | 109 | |
| AK101896.1 | - | 6 - 7 | 2403 | 109 | ||
| EU837261.1 | - | 8 - 9 | 1129 | 109 | ||
More Information
Set Membership: |
|

LOC4347359
Genomic Map