Assay Details
| Gene Symbol: LOC4345839 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015795151.2 | XP_015650637.1 | 4 - 5 | 547 | 94 | |
| GenBank mRNA | AB110175.1 | - | 4 - 5 | 449 | 94 | |
| AK068050.1 | - | 4 - 5 | 517 | 94 | ||
More Information
Set Membership: |
|

LOC4345839
Genomic Map