Assay Details
| Gene Symbol: LOC4345899 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015795006.2 | XP_015650492.1 | 2 - 3 | 307 | 163 | |
| GenBank mRNA | AK100183.1 | - | 2 - 3 | 292 | 163 | |
| HQ858865.1 | - | 1 - 2 | 137 | 163 | ||
More Information
Set Membership: |
|

LOC4345899
Genomic Map