Assay Details
| Gene Symbol: LOC4345899 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015795004.2 | XP_015650490.2 | 6 - 7 | 897 | 89 | |
| XM_015795006.2 | XP_015650492.1 | 7 - 8 | 1099 | 89 | ||
| GenBank mRNA | AK070237.1 | - | 6 - 7 | 822 | 89 | |
| AK100183.1 | - | 7 - 8 | 1084 | 89 | ||
| HQ858865.1 | - | 6 - 7 | 929 | 89 | ||
More Information
Set Membership: |
|

LOC4345899
Genomic Map