Assay Details
| Gene Symbol: LOC4346660 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015756500.1 | XP_015611986.1 | 4 - 5 | 678 | 69 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC4346660| Gene Symbol: LOC4346660 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015756500.1 | XP_015611986.1 | 4 - 5 | 678 | 69 | |
Set Membership: |
|