Assay Details
| Gene Symbol: LOC4346713 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015755380.2 | XP_015610866.1 | 1 - 2 | 165 | 101 | |
| GenBank mRNA | AK073821.1 | - | 1 - 2 | 142 | 101 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015755380.2 transcripts.
Set Membership: |
|

LOC4346713
Genomic Map