Assay Details
| Gene Symbol: LOC4346713 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015755379.2 | XP_015610865.1 | 3 - 4 | 388 | 83 | |
| XM_015755380.2 | XP_015610866.1 | 3 - 4 | 382 | 83 | ||
| GenBank mRNA | AK073821.1 | - | 3 - 4 | 359 | 83 | |
More Information
Set Membership: |
|

LOC4346713
Genomic Map