Assay Details
| Gene Symbol: LOC4346871 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015756530.2 | XP_015612016.1 | 1 - 2 | 137 | 130 | |
| XM_015756531.2 | XP_015612017.1 | 1 - 2 | 137 | 130 | ||
| GenBank mRNA | AK120754.1 | - | 1 - 2 | 98 | 130 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015756530.2, XM_015756531.2 transcripts.
Set Membership: |
|

LOC4346871
Genomic Map