Assay Details
| Gene Symbol: LOC4346871 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015756530.2 | XP_015612016.1 | 2 - 3 | 220 | 72 | |
| GenBank mRNA | AB110222.1 | - | 2 - 3 | 168 | 72 | |
| AK120754.1 | - | 2 - 3 | 181 | 72 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015756530.2 transcripts.
Set Membership: |
|

LOC4346871
Genomic Map