Assay Details
| Gene Symbol: LOC4346871 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015756530.2 | XP_015612016.1 | 3 - 4 | 271 | 61 | |
| XM_015756531.2 | XP_015612017.1 | 2 - 3 | 2526 | 61 | ||
| GenBank mRNA | AB110222.1 | - | 3 - 4 | 219 | 61 | |
| AK072157.1 | - | 2 - 3 | 2455 | 61 | ||
| AK120754.1 | - | 3 - 4 | 232 | 61 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015756530.2, XM_015756531.2 transcripts.
Set Membership: |
|

LOC4346871
Genomic Map