Assay Details
| Gene Symbol: LOC4347066 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015756333.2 | XP_015611819.1 | 1 | 2963 | 119 | |
| GenBank mRNA | AK065377.1 | - | 1 | 2953 | 119 | |
More Information
Set Membership: |
|

LOC4347066
Genomic Map