Assay Details
Gene Symbol: LOC4347086 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015756510.2 | XP_015611996.1 | 3 - 4 | 1636 | 107 | |
GenBank mRNA | AK065475.1 | - | 3 - 4 | 1637 | 107 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
