Assay Details
| Gene Symbol: LOC4347226 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015756315.1 | XP_015611801.1 | 4 - 5 | 731 | 135 | |
| GenBank mRNA | AK062254.1 | - | 4 - 5 | 682 | 135 | |
| AK072941.1 | - | 4 - 5 | 737 | 135 | ||
More Information
Set Membership: |
|

LOC4347226
Genomic Map