Assay Details
| Gene Symbol: LOC4347226 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015756315.1 | XP_015611801.1 | 6 - 7 | 1010 | 69 | |
| GenBank mRNA | AK062254.1 | - | 6 - 7 | 961 | 69 | |
More Information
Set Membership: |
|

LOC4347226
Genomic Map