Assay Details
| Gene Symbol: LOC4347482 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015756789.2 | XP_015612275.1 | 1 - 2 | 156 | 108 | |
| GenBank mRNA | AK066999.1 | - | 1 - 2 | 162 | 108 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015756789.2 transcripts.
Set Membership: |
|

LOC4347482
Genomic Map