Assay Details
| Gene Symbol: LOC4347482 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015756789.2 | XP_015612275.1 | 7 - 8 | 920 | 148 | |
| GenBank mRNA | AK066999.1 | - | 7 - 8 | 926 | 148 | |
More Information
Set Membership: |
|

LOC4347482
Genomic Map